Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Mercaptoethanol
2-Mercaptoethanol ≥98%
2-Mercaptoethanol ≥98%
:
Thermo Fisher Scientific
2-Mercaptoethanol ≥98%
:
Thermo Fisher Scientific
:
60-24-2
:
:
HSCH₂CH₂OH
:
MFCD00004890
:
14,5869
:
156…158 °C
:
–100 °C
:
1.114
:
73 °C (163 °F)
:
Ambient
:
2966
:
6.1,II
:
78.14 g/mol
:
200-464-6
:
GHS06
:
GHS05
:
GHS07
msds_ALFAA15890
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
VWR®, Transfer Pipettes, Graduated
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR® Disposable Microcentrifuge Tubes
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals