Order Entry
France
Orders LinkContactUsLinkComponent
2-Propanol

2-Propanol

10 L
Catalog # PRLS87561.410
Flammable
Irritant
Supplier:   PROLABO CMP
2-Propanol
Catalog # PRLS87561.410
Supplier:  PROLABO CMP
CAS Number:  67-63-0
2-Propanol
2-Propanol

10 L

CAS Number:  67-63-0
Price
203,00 €
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0.786 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25
Specifications

Specifications

  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.