
Stearic anhydride 98%
Catalog # ACRO419710250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to yellow to light beige Powder or flakes
- Infrared spectrum:Conforms
- Titration excess NaOH:≥97.5 %
- GC:≥96.0 %
Specification Test Results
About this item
:
:550.95 g/mol
Melting Pt:69…71 °C
Density:0.84 g/cm³
EINECS:211-318-6
:GHS07
Storage Temperature:Freezer
CAS Number:638-08-4
Formula:C₃₆H₇₀O₃
MDL Number:MFCD00009004