


2-(Dimethylamino)-ethanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-01-0
:
Formula:C₄H₁₁NO
MDL Number:MFCD00002846
:89.14 g/mol
Merck Index:13,02863
Boiling Pt:132…135 °C (1013 hPa)
Melting Pt:–59 °C
Density:0.8871 g/cm³ (20 °C)
EINECS:203-542-8
UN:2051
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
Flash Pt:39 °C
:GHS02
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-59 °C
- Molar Mass:89.14 g/mol
- Boiling Point:132 - 135 °C (1013 hPa)
- Vapor Pressure:6.12 hPa (20 °C)
- Flash Point:39 °C DIN 51755 Part 1
- Refractive Index:1.4294 (20 °C, 589 nm)
- Explosion Limit:1.4 - 12.2 %(V)
- Density:0.89 g/cm3 (20 °C)
- pH:10.5 - 11 (100 g/l, H2O, 20 °C)
- Ignition Point:245 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.03237.0100_TDS_EN ProductPage_803237
Product specification
msds_803237
SDS
Frequently Bought Together