

Methyl formate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-31-3
:
Formula:C₂H₄O₂
MDL Number:MFCD00003291
:60.05 g/mol
Merck Index:12,06153
Boiling Pt:32 °C (1013 hPa)
Melting Pt:–100.4 °C
Density:0.9713 g/cm³ (20 °C)
EINECS:203-481-7
UN:1243
ADR:3,I
:M-Clarity™ quality level = MQ200
Flash Pt:–32 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:300 g/l (20 °C)
- Melting Point:-100.4 °C
- Molar Mass:60.05 g/mol
- Boiling Point:31.5 °C (1013 hPa)
- Vapor Pressure:644 hPa (20 °C)
- Flash Point:-32 °C
- Refractive Index:1.3433 (20 °C, 589 nm)
- Explosion Limit:5 - 23 %(V)
- Density:0.968 g/cm3 (20 °C)
- pH:4 - 5 (200 g/l, H2O, 20 °C)
- Ignition Point:450 °C
Specification Test Results
Frequently Bought Together