Order Entry
France
ContactUsLinkComponent
 
 
Methyl hexanoate for synthesis, Sigma-Aldrich®
 :  Merck
 
 
Methyl hexanoate for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    1.33 g/l (20 °C)
  • Melting Point:
    -71 °C
  • Molar Mass:
    130.19 g/mol
  • Boiling Point:
    151 °C (1013 hPa)
  • Vapor Pressure:
    3.7 hPa (20 °C)
  • Flash Point:
    41 °C
  • Refractive Index:
    1.396 (20 °C, 589 nm)
  • Density:
    0.88 g/cm3 (20 °C)

 

Frequently Bought Together