Order Entry
France
ContactUsLinkComponent
N,N-Dimethylacetamide 99.8+% for HPLC
Irritant
Health
N,N-Dimethylacetamide 99.8+% for HPLC
Certificates
undefined
Irritant
Health
N,N-Dimethylacetamide 99.8+% for HPLC

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • GC:
    ≥99.80 %
  • UV:
    1 cm cell vs HPLC-grade water
  • Non-volatile matter:
    ≤0.0003 %
  • Acidity:
    ≤0.0005 meq/g
  • Alkalinity:
    ≤0.003 meq/g

Specification Test Results

Related Information

Frequently Bought Together