


Chloroacetic anhydride 97%
:541-88-8
:
:C₄H₄Cl₂O₃
:MFCD00000929
:170.98 g/mol
:13,02130
:203 °C (1013 hPa)
:51…59 °C
:1,5497 g/cm³ (20 °C)
:143 °C
:Ambient
:208-794-2
:2923
:8,II
:GHS05
:GHS06
:GHS09
- Appearance:White to brown Powder or crystals and/or chunks or fused solid
- Infrared spectrum:Conforms
- Melting point:50 to 60 °C
- GC:≥96.0 %
Frequently Bought Together