Order Entry
France
ContactUsLinkComponent
Chloroacetic acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
Chloroacetic acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 20245.238
Supplier:  VWR Chemicals
CAS Number:  
undefined
Chloroacetic acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 20245.238
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Melting point:
    61 - 63 °C
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.05 %
  • Water:
    Max. 0.5 %
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :94.5 g/mol
Merck Index:13,02129
Boiling Pt:188 °C (1013 hPa)
Melting Pt:56…57 °C
Density:1.4043 g/cm³ (20 °C)
Flash Pt:126 °C
EINECS:201-178-4
UN:1751
ADR:6.1 (8),II
 :GHS06
 :GHS05
 :GHS09
Storage Temperature:Ambient
CAS Number:79-11-8
Formula:ClCH₂COOH
MDL Number:MFCD00002683