Chloroacetic acid
Catalog # EHERC11348500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:94.5 g/mol
Merck Index:13,02129
Boiling Pt:188 °C (1013 hPa)
Melting Pt:56…57 °C
Density:1.4043 g/cm³ (20 °C)
Flash Pt:126 °C
EINECS:201-178-4
UN:1751
ADR:6.1,II
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
CAS Number:79-11-8
Formula:ClCH₂COOH
MDL Number:MFCD00002683