Chloroacetic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-11-8
:
Formula:ClCH₂COOH
MDL Number:MFCD00002683
:94.5 g/mol
Merck Index:14,2112
Boiling Pt:189 °C
Melting Pt:60…63 °C
Density:1.404
Flash Pt:126 °C (258 °F)
Storage Temperature:Ambient
EINECS:201-178-4
UN:1751
ADR:6.1,II
:GHS06
:GHS05
:GHS09
Related Information
Frequently Bought Together