Chloroacetic acid 99%, flakes
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-11-8
:
Formula:ClCH₂COOH
MDL Number:MFCD00002683
:94.5 g/mol
Merck Index:13,02129
Boiling Pt:188 °C (1013 hPa)
Melting Pt:61…63 °C
Density:1.58 g/cm³
Flash Pt:126 °C
Storage Temperature:Ambient
EINECS:201-178-4
UN:1751
ADR:6.1,II
:GHS06
:GHS05
:GHS09
Specification Test Results
- Appearance:White Crystalline flakes
- Infrared spectrum:Conforms
- Melting point:61 to 63 °C
- Water:≤0.3 %
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together