Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61 °C (1.013 hPa)
Melting Pt:–61.3 °C
Density:1.485 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:Stabilised with amylene
EINECS:200-663-8
REACH:05-2114614906-45
:GHS07
:GHS08
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.80 %
- 2-methyl-2-buten (GC):20 - 200 ppm
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.03 %
- Free chlorine:complying
- Lead (Pb):Max. 0.05 ppm
- Acetone, aldehydes (neßler test):complying
- Free acid and chloride:complying
- Reaction against H₂SO₄:complying
- APHA:Max. 10
- Contains:amylenes as stabilizer
Specification Test Results
Frequently Bought Together