Order Entry
France
ContactUsLinkComponent
Chloroform, EMPARTA® ACS analytical reagent, Supelco®
Chloroform, EMPARTA® ACS analytical reagent, Supelco®
Supplier:  Merck
Certificates
undefined
Chloroform, EMPARTA® ACS analytical reagent, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    8 g/l (20 °C)
  • Melting Point:
    -63 °C
  • Molar Mass:
    119.38 g/mol
  • Boiling Point:
    61 °C (1013 hPa)
  • Vapor Pressure:
    211 hPa (20 °C)
  • Refractive Index:
    1.45
  • Density:
    1.48 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together