Supelco® Organic Reference Standard, 1,1,2,2-Tetrachloroethane
Catalog # 46259-5ML
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:79-34-5
- Description:1,1,2,2-Tetrachloroethane
- Storage conditions:2…8 °C
- Hazard class description:H310 + H330 - H411
- Shipping conditions:STD
- MDL No.:MFCD00000848
- M-Clarity quality level:MQ100
- Synonym(s):Acetylene tetrachloride
- Melting point:–43 °C (lit.)
- Formula:CHCl₂CHCl₂
- Molecular weight:167,85
- Density:1,586 g/ml at 25 °C (lit.)
- Boiling point:147 °C (lit.)
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310