Seignette's salt solution (potassium sodium tartrate tetrahydrate and Nessler reagent)
Supplier: PROLABO CMP
Certificates
About this item
:
Merck Index:13,07755
Melting Pt:70…80 °C
Density:∼1.767 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-156-8
Frequently Bought Together