Order Entry
France
ContactUsLinkComponent
 
Organic reference standard, 2,4,4',6-Tetrachlorobiphenyl (PCB No. 75) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-75
 :  
Organic reference standard, 2,4,4',6-Tetrachlorobiphenyl (PCB No. 75) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-75
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    32598-12-2
  • Description:
    2,4,4',6-Tetrachlorobiphenyl (PCB No. 75) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    2,4,4',6-Tetrachlorobiphenyl (PCB No. 75)