L(+)-Aspartic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-84-8
:
Formula:C₄H₇NO₄
MDL Number:MFCD00002616
:133.1 g/mol
Merck Index:14,840
Boiling Pt:311 °C (1013 hPa)
Melting Pt:>300 °C (dec.)
Density:1.66
Storage Temperature:Ambient
EINECS:200-291-6
Related Information
Frequently Bought Together