Organic reference standard, 2,2',5-Trichlorobiphenyl (PCB No. 18) 100 µg/ml in isooctane, CIL
PROOCIL-PCB-18-CS
:
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:37680-65-2
- Description:2,2',5-Trichlorobiphenyl (PCB No. 18) 100 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,2',5-Trichlorobiphenyl (PCB No. 18)