Order Entry
France
ContactUsLinkComponent
L(+)-Sodium lactate
L(+)-Sodium lactate
Catalog # A1004.0100
Supplier:  PanReac AppliChem
CAS Number:  
L(+)-Sodium lactate
Catalog # A1004.0100
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :112.06 g/mol
EINECS:212-762-3
CAS Number:867-56-1
Formula:C₃H₅NaO₃
MDL Number:MFCD00066576