Order Entry
France
ContactUsLinkComponent
L-Arginine hydrochloride
L-Arginine hydrochloride
Catalog # PROOMM0569.00
CAS Number:  
L-Arginine hydrochloride
Catalog # PROOMM0569.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :210.66 g/mol
Merck Index:13,00785
Melting Pt:218 °C
Density:1.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-275-1
CAS Number:1119-34-2
Formula:C₆H₁₅ClN₄O₂
MDL Number:MFCD00064550