D-(-)-Diethyl tartrate (made from unnatural tartaric acid) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (2S,3S)-Diethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Diethyl tartrate, (2S,3S)-(-)-Diethyl tartrate, (-)-Diethyl D-tartrate, (S,S)-(-)-Tartaric acid diethyl ester, D(-)-Tartaric acid diethyl ester
CAS Number:13811-71-7
:
Formula:C₈H₁₄O₆
MDL Number:MFCD00064451
Boiling Pt:280 °C (760 mmHg)
Melting Pt:17 °C
Density:1.2 g/cm³
Molecular Weight:206.2 g/mol
Flash Pt:93 °C
Storage Temperature:Ambient
EINECS:237-458-8
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4450 to 1.4470 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:-7.5° to -9° (20°C, 589 nm) (neat)
Specification Test Results
Frequently Bought Together