Order Entry
France
ContactUsLinkComponent
 
Organic reference standard, trans-Dichloroethylene 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA12422600ME
 :  
Organic reference standard, trans-Dichloroethylene 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA12422600ME
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    156-60-5
  • Description:
    trans-Dichloroethylene 100 µg/ml in methanol
  • Formula:
    C₂H₂Cl₂
  • Description:
    trans-1,2-Dichloroethene