Order Entry
France
ContactUsLinkComponent
Organic reference standard, 1,1-Dichloroethylene 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 1,1-Dichloroethylene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12422400ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 1,1-Dichloroethylene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12422400ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    75-35-4
  • Description:
    1,1-Dichloroethylene 100 µg/ml in methanol
  • Formula:
    C₂H₂Cl₂ 
  • Description:
    1,1-Dichloroethylene

Specifications

About this item