Order Entry
France
ContactUsLinkComponent
 
Organic reference standard, Bromodichloromethane 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA10726700ME
 :  
Organic reference standard, Bromodichloromethane 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA10726700ME
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    75-27-4
  • Description:
    Bromodichloromethane 100 µg/ml in methanol
  • Formula:
    CHBrCl₂
  • Description:
    Bromodichloromethane