Cobalt(II) nitrate hexahydrate 99-102%, AnalaR NORMAPUR® analytical reagent nickel-free
22913.297
:
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
- Assay:99 - 102 %
- Insolubility in water:Max. 50 ppm
- Cl (Chloride):Max. 10 ppm
- NH4 (Ammonium):Max. 0.05 %
- SO4 (Sulphate):Max. 50 ppm
- Ni (Nickel):Max. 50 ppm
- Pb (Lead):Max. 10 ppm
- Zn (Zinc):Max. 100 ppm
:
:291.03 g/mol
:13,02469
:57 °C
:1.88 g/cm³ (20 °C)
:Ambient
:233-402-1
:1477
:5.1,III
:GHS08
:GHS09
:10026-22-9
:Co(NO₃)₂·6H₂O
:MFCD00149647