Order Entry
France
ContactUsLinkComponent
Nickel(II) chloride hexahydrate ≥97%, GPR RECTAPUR®
Health
Toxic
Environment
Nickel(II) chloride hexahydrate ≥97%, GPR RECTAPUR®
Catalog # 25850.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Toxic
Environment
Nickel(II) chloride hexahydrate ≥97%, GPR RECTAPUR®
Catalog # 25850.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 97 %
  • SO4 (Sulphate):
    Max. 0.1 %
  • Cd (Cadmium):
    Max. 20 ppm
  • Cu (Copper):
    Max. 20 ppm
  • Fe (Iron):
    Max. 100 ppm
  • Pb (Lead):
    Max. 20 ppm
  • Zn (Zinc):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :237.69 g/mol
Merck Index:13,06529
Melting Pt:140 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-743-0
UN:3288
ADR:6.1,III
 :GHS08
 :GHS06
 :GHS09
CAS Number:7791-20-0
Formula:NiCl₂·6H₂O
MDL Number:MFCD00149809