Order Entry
France
ContactUsLinkComponent
 
Health
Toxic
Environment
Cadmium(II) chloride hemipentahydrate 79.5-81.0% ACS, Fluka™
Catalog # 239208-500G
CAS Number:  
Health
Toxic
Environment
Cadmium(II) chloride hemipentahydrate 79.5-81.0% ACS, Fluka™
Catalog # 239208-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Vapor Pressure:
    10 mmHg ( 656 °C)
  • Assay of CdCl₂:
    79.5 - 81.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • Calcium (Ca):
    Max. 0.005 %
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 0.02 %
  • Sodium (Na):
    Max. 0.05 %
  • Lead (Pb):
    Max. 0.005 %
  • Zinc (Zn):
    Max. 0.05 %
  • Ammonium (NH₄):
    Max. 0.005 %
  • Nitrate, nitrite (as NO₃):
    Max. 0.003 %
  • Sulfate (SO₄):
    Max. 0.005 %

Specification Test Results

About this item

 :
 :228.35 g/mol
Merck Index:13,01617
Melting Pt:568 °C
Density:3.327 g/cm³
Storage Temperature:Ambient
EINECS:233-296-7
UN:2570
ADR:6.1,III
 :GHS08
 :GHS06
 :GHS09
CAS Number:7790-78-5
Formula:CdCl₂·2.5H₂O
MDL Number:MFCD00799071