Order Entry
France
ContactUsLinkComponent
Bole white (kaolin), powder, EMPROVE® EXPERT Ph. Eur., BP suitable for use as excipient, for internal use low bacterial content, SAFC®
Bole white (kaolin), powder, EMPROVE® EXPERT Ph. Eur., BP suitable for use as excipient, for internal use low bacterial content, SAFC®
Catalog # 1.05006.9025
CAS Number:  
Bole white (kaolin), powder, EMPROVE® EXPERT Ph. Eur., BP suitable for use as excipient, for internal use low bacterial content, SAFC®
Catalog # 1.05006.9025
CAS Number:  

Specifications

  • Pack type:
    Cardboard carton
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :258.16 g/mol
Merck Index:13,05300
Melting Pt:1760 °C
Density:0.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:310-194-1
REACH:exempt
CAS Number:1332-58-7
Formula:Al₂SiO₇·2H₂O
MDL Number:MFCD00062311