Strontium hydroxide octahydrate ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1311-10-0
:
Formula:H₂O₂Sr·8H₂O
MDL Number:MFCD00150395
:265.76 g/mol
Merck Index:14,8843
Melting Pt:100 °C (-8H₂O)
Density:1.900
Storage Temperature:Ambient
EINECS:242-367-1
UN:3262
ADR:8,III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together