N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1465-25-4
:
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556
:259.18 g/mol
Merck Index:12,06496
Melting Pt:188…190 °C
:M-Clarity™ quality level = MQ200
EINECS:215-981-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together