Hydrazine dihydrochloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Hydrazine Dihydrochloride, Hydrazinium dichloride
CAS Number:5341-61-7
:
Formula:H₆Cl₂N₂
MDL Number:MFCD00064543
:104.97 g/mol
Merck Index:13,04789
Melting Pt:189 °C
Density:1.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-283-2
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:3288
:GHS06
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:2700 g/l (20 °C)
- Melting Point:198 °C (decomposition)
- Molar Mass:104.97 g/mol
- Bulk Density:770 kg/m3
- Density:1.42 g/cm3 (20 °C)
- pH:0.5 (50 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together