Hydroxylammonium chloride, TECHNICAL
Catalog # 24706.238
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:69.49 g/mol
Merck Index:13,04853
Boiling Pt:306 °C (1013 hPa)
Melting Pt:151 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-798-2
UN:2923
ADR:8 (6.1),III
:GHS08
:GHS07
:GHS05
:GHS09
CAS Number:5470-11-1
Formula:NH₂OH·HCl
MDL Number:MFCD00051089