Basic fuchsin in ACS Reagent grade water ≥85% (dye content), Sigma-Aldrich®
215597-25G
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
- Appearance (Colour):Green
- Appearance (Form):Powder or Crystals
- Infrared spectrum:Conforms to Structure
- Wavelength
c = 0.003 g/L in 50% Ethanol:543 - 549 nm - Extinction Coefficient:≥80000
- Wavelength:286 - 292 nm
- Carbon:59.9 - 81.0%
- Nitrogen:11.0 - 14.9%
- Purity (HPLC):≥85.0%
: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
:
:337.85 g/mol
:211-189-6
:GHS08
:Ambient
:632-99-5
:58969-01-0
:C₂₀H₂₀ClN₃
:MFCD00012569
:235
:1.22 g/cm³ (20 °C)
:12,05686
:M-Clarity™ quality level = MQ100