Iron(III) chloride 55% in aqueous solution
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:7705-08-0
:
Formula:FeCl₃
MDL Number:MFCD00011005
:162.21 g/mol
EINECS:231-729-4
UN:2582
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together