Order Entry
France
ContactUsLinkComponent
Ammonium thiocyanate 20 g/l in aqueous solution
Ammonium thiocyanate 20 g/l in aqueous solution
Catalog # PRLS85562.410
Supplier:  PROLABO CMP
CAS Number:  
Ammonium thiocyanate 20 g/l in aqueous solution
Catalog # PRLS85562.410
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :76.12 g/mol
Storage Temperature:Ambient
EINECS:217-175-6
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428