Order Entry
France
Orders LinkContactUsLinkComponent
Ammonia solution 30%

Ammonia solution 30%

Plastic bottle, 2 L
Catalog # PRLS85482.300
Corrosive
Irritant
Environment
Supplier:   PROLABO CMP
Ammonia solution 30%
Catalog # PRLS85482.300
Supplier:  PROLABO CMP
CAS Number:  1336-21-6
Ammonia solution 30%

Plastic bottle, 2 L

CAS Number:  1336-21-6
Price
60,60 €
Log in for availability
1 * 2 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.