PCR/gel clean-up kits
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
PCR/Gel Clean Up Kits are quick and simple kits for purifying DNA from PCR, gels and labelling reactions for a variety of downstream applications.
- High recoveries for fragments down to 50 bp
- Small elution volume down to 15 µl
- Binding buffer with pH indicator
- Validated for use with CGH labelling kit
- Less preparation time: 10 mins for 6 preparations
Clean-up columns use silica membrane technology in a mini spin column format to deliver easy and efficient DNA purification from PCR products, agarose gels, and labelling reactions.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Amplified/modified DNA
- Target molecule:DNA