These high wattage, constant-temperature dry block heaters are economical, versatile and compact. The multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures.
- PID temperature controller maintains precise temperature control
- Samples are heated to temperature quickly and accurately
- Has the temperature uniformity of ±0,1 °C at 37 °C within the block
- Can be used for coagulation and RH studies
Each model accepts separate interchangeable modular blocks, accommodating various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes. Each block has a thermometer well for measuring block temperature. Anodised aluminum modular blocks provide superior temperature stability and heat transfer.
Dual temperature control knobs with dial markings from 1 to 10 for low temperature and high temperature adjustments. Low range knob adjusts from ambient to 100 °C and high range knob adjusts from 75 to 150 °C.
Units can be run in conditions from 18 to 33 °C (64 to 91 °F), 20 to 80% relative humidity, non-condensing.