Order Entry
France
ContactUsLinkComponent
Tetraethylammonium hydroxide 40% in aqueous solution
Corrosive
Tetraethylammonium hydroxide 40% in aqueous solution
Catalog # MOLEM86474667
Supplier:  Molekula
CAS Number:  
Corrosive
Tetraethylammonium hydroxide 40% in aqueous solution
Catalog # MOLEM86474667
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :147.26 g/mol
EINECS:201-073-3
UN:3267
ADR:8,II
 :GHS05
CAS Number:77-98-5
Formula:C₈H₂₁NO
MDL Number:MFCD00009024