
Barium chloride 10% in aqueous solution
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:10361-37-2
:
Formula:BaCl₂
MDL Number:MFCD00003445
:208.24 g/mol
Boiling Pt:∼100 °C (1013 hPa)
Storage Temperature:Ambient
EINECS:233-788-1
UN:3287
ADR:6.1,III
:GHS07
Frequently Bought Together