Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium thiocyanate
Potassium thiocyanate 10% in aqueous solution
Potassium thiocyanate 10% in aqueous solution
PRLSMC306755.0100
:
PROLABO CMP
:
Potassium thiocyanate 10% in aqueous solution
PRLSMC306755.0100
:
PROLABO CMP
:
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
:
:
97.18 g/mol
:
Ambient
:
206-370-1
:
333-20-0
:
KSCN
:
MFCD00011413