Order Entry
France
Orders LinkContactUsLinkComponent
 

Sodium hydroxide 10% in aqueous solution

Supplier:   PROLABO CMP
Sodium hydroxide 10% in aqueous solution
Supplier:  PROLABO CMP
Certificates
Sodium hydroxide 10% in aqueous solution

3 Options Available Below

Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II

 

Product Family Options

Product Information