Potassium hydroxide 38% in aqueous solution, GPR RECTAPUR®
Plastic bottle, 5 L
Supplier: VWR Chemicals
Potassium hydroxide 38% in aqueous solution, GPR RECTAPUR®
Catalog # 26629.361
Supplier: VWR Chemicals
CAS Number: 1310-58-3
Jump to:
Product Details & Documents
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Storage Temperature:Ambient
- EINECS:215-181-3
- UN:1814
- ADR:8,II
Documents
Specifications
Specification Test Results
- Assay:38 - 40 %
- Density (20/4):1.374 - 1.396
- Heavy metals (as Pb):Max. 20 ppm
- Cl (Chloride):Max. 100 ppm
- Fe (Iron):Max. 20 ppm
- SO4 (Sulphate):Max. 100 ppm
Specifications
- Pack type:Plastic bottle
- Pk:5 L



