Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium iodide
Potassium iodide 40 g/l in aqueous solution
Potassium iodide 40 g/l in aqueous solution
PRLSMC307226.0500
:
PROLABO CMP
:
Potassium iodide 40 g/l in aqueous solution
PRLSMC307226.0500
:
PROLABO CMP
:
Pack type:
Glass bottle
Pk:
500 mL
Seq:
2725
:
:
166 g/mol
:
Ambient
:
231-659-4
:
7681-11-0
:
Kl
:
MFCD00011405