
Potassium iodide 4% in aqueous solution
Catalog # PRLSMC240319.5000
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:166 g/mol
EINECS:231-659-4
:GHS08
Storage Temperature:Ambient
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405