Order Entry
France
ContactUsLinkComponent
 
Iodine 0.01 mol/l (0.02 N) in aqueous solution
Catalog # PRLS85123.320
Supplier:  PROLABO CMP
CAS Number:  
Iodine 0.01 mol/l (0.02 N) in aqueous solution
Catalog # PRLS85123.320
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

About this item

 :
 :253.81 g/mol
Storage Temperature:Ambient
EINECS:231-442-4
UN:3264
ADR:8,II
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355