Order Entry
France
ContactUsLinkComponent
Ammonium thiocyanate 0.02 mol/l (0.02 N) in aqueous solution
Ammonium thiocyanate 0.02 mol/l (0.02 N) in aqueous solution
Catalog # PRLSMC302634.0500
Supplier:  PROLABO CMP
CAS Number:  
Ammonium thiocyanate 0.02 mol/l (0.02 N) in aqueous solution
Catalog # PRLSMC302634.0500
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :76.12 g/mol
Storage Temperature:Ambient
EINECS:217-175-6
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428