Potassium thiocyanate 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31480.292
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Titer (20°C;real value 0.2 % accuracy):0.998 - 1.002 mol/l
- Appearance:Clear colourless liquid
Specification Test Results
About this item
:
:97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413