
Sodium hydroxide 1/1.4 mol/l (1/1.4 N) in aqueous solution
Catalog # PRLSMC307901.5000
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:40 g/mol
Density:1.04 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:3266
ADR:8,III
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548